APOH11A9C3A2 Apollo Scientific Ltd

4-Hydroxynonafluorobiphenyl

  • MDL number MFCD00134468
  • Cas Number : 2894-87-3
  • Molecular Weight : 332.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2',3,3',4',5,5',6,6'-nonafluoro[1,1'-biphenyl]-4-ol
Inchi Code 1S/C12HF9O/c13-3-1(4(14)8(18)9(19)7(3)17)2-5(15)10(20)12(22)11(21)6(2)16/h22H
Inchi Key MLXJPZBMOLLESC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxynonafluorobiphenyl

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/4-hydroxynonafluorobiphenyl
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart