APOH11A9C4EC Apollo Scientific Ltd

2,6-Difluoro-4-hydrazinopyridine

  • MDL number MFCD08436095
  • Cas Number : 837364-94-0
  • Molecular Weight : 145.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-difluoro-4-hydrazinopyridine
Inchi Code 1S/C5H5F2N3/c6-4-1-3(10-8)2-5(7)9-4/h1-2H,8H2,(H,9,10)
Inchi Key YDUYJJIXZAXMIJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-4-hydrazinopyridine

Safety Information

signalword Danger
pcodes P280;P301+P310;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-4-hydrazinopyridine
pictograms GHS05;GHS06
hstatements H301;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart