APOH11A9C500 Apollo Scientific Ltd

3,4-Difluorophenyl isothiocyanate

  • MDL number MFCD00175109
  • Cas Number : 113028-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.35
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 170
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10615

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10615
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart