APOH11A9C550 Apollo Scientific Ltd

2-Hydrazino-4-(trifluoromethyl)pyridine

  • MDL number MFCD08056340
  • Cas Number : 89570-84-3
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydrazino-4-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-hydrazino-4-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart