APOH11A9C5B9 Apollo Scientific Ltd

3-Chloro-2-iodo-5-(trifluoromethyl)pyridine

  • MDL number MFCD09264543
  • Cas Number : 134161-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36921

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=36921
Availability details for
Quantity
  • Availability
  • Add to Cart