APOH11A9C63E Apollo Scientific Ltd

5-Chloro-3-fluoropyridin-2ylhydrazine

  • MDL number MFCD09972183
  • Cas Number : 248255-70-1
  • Molecular Weight : 161.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-fluoro-2-hydrazinopyridine
Inchi Code 1S/C5H5ClFN3/c6-3-1-4(7)5(10-8)9-2-3/h1-2H,8H2,(H,9,10)
Inchi Key TZBBCECFLONWDG-UHFFFAOYSA-N

Description

Synonym 5-Chloro-3-fluoro-2-hydrazinopyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-3-fluoropyridin-2ylhydrazine

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/5-chloro-3-fluoropyridin-2ylhydrazine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart