APOH11A9C68F Apollo Scientific Ltd

2-(3,4-Dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol

  • MDL number MFCD03412228
  • Cas Number : 2379-17-1
  • Molecular Weight : 272.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
Iupac Name 2-(3,4-dimethylphenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
Inchi Code 1S/C11H10F6O/c1-6-3-4-8(5-7(6)2)9(18,10(12,13)14)11(15,16)17/h3-5,18H,1-2H3
Inchi Key DDLQAOJQQSJPIH-UHFFFAOYSA-N

Description

Melting Point >110
Synonym 4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)-o-xylene, 1,2-Dimethyl-4-[perfluoro(2-hydroxyprop-2-yl)]benzene
Flash Point 101-102/20mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-34-dimethylphenyl-111333-hexafluoropropan-2-ol

Safety Information

signalword Warning
pcodes P302+P352;P305+P351+P338;P362+P364
MSDS https://store.apolloscientific.co.uk/product/2-34-dimethylphenyl-111333-hexafluoropropan-2-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart