APOH11A9C719 Apollo Scientific Ltd

2-Bromo-4-fluoro-5-(trifluoromethyl)aniline

  • MDL number MFCD04973755
  • Cas Number : 193090-60-7
  • Molecular Weight : 258.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 24-29
Iupac Name 2-bromo-4-fluoro-5-(trifluoromethyl)aniline
Inchi Code 1S/C7H4BrF4N/c8-4-2-5(9)3(1-6(4)13)7(10,11)12/h1-2H,13H2
Inchi Key ZRMRCSMDONHRKT-UHFFFAOYSA-N

Description

Synonym 5-Amino-4-bromo-2-fluorobenzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 24-29
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-5-trifluoromethylaniline

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-5-trifluoromethylaniline
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart