APOH11A9C838 Apollo Scientific Ltd

3-Amino-6-(trifluoromethyl)-1H-indazole

  • MDL number MFCD03426695
  • Cas Number : 2250-55-7
  • Molecular Weight : 201.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-165
Synonym 6-(Trifluoromethyl)-1H-indazol-3-amine
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-6-trifluoromethyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-6-trifluoromethyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart