APOH11A9CA2E Apollo Scientific Ltd

3,5-Dichloro-4-fluoroaniline

  • MDL number MFCD09835492
  • Cas Number : 2729-34-2
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 102
Iupac Name 3,5-dichloro-4-fluoroaniline
Inchi Code 1S/C6H4Cl2FN/c7-4-1-3(10)2-5(8)6(4)9/h1-2H,10H2
Inchi Key NKTZSKQMAITPBQ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 102
Product Link https://store.apolloscientific.co.uk/product/35-dichloro-4-fluoroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloro-4-fluoroaniline
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart