APOH11A9CA39 Apollo Scientific Ltd

4-Bromo-2-chloro-6-fluoroiodobenzene

  • MDL number MFCD09878165
  • Cas Number : 83027-73-0
  • Molecular Weight : 335.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-1-chloro-3-fluoro-2-iodobenzene
Inchi Code 1S/C6H2BrClFI/c7-3-1-4(8)6(10)5(9)2-3/h1-2H
Inchi Key WQAABVMKMRFARY-UHFFFAOYSA-N

Description

Synonym 5-Bromo-1-chloro-3-fluoro-2-iodobenzene
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-chloro-6-fluoroiodobenzene

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-chloro-6-fluoroiodobenzene
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart