APOH11A9CA4F Apollo Scientific Ltd

Methyl 2-fluoro-5-hydroxybenzoate

  • MDL number MFCD12406843
  • Cas Number : 1084801-91-1
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Carboxy-4-fluorophenol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-fluoro-5-hydroxybenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-fluoro-5-hydroxybenzoate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart