APOH11A9CA6A Apollo Scientific Ltd

(2Z)-3-(Diethylamino)-2-fluoroprop-2-enal

  • MDL number MFCD18633001
  • Cas Number : 152873-64-8
  • Molecular Weight : 145.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2Z)-3-(Diethylamino)-2-fluoroacrolein, (2Z)-3-(Diethylamino)-2-fluoroacrylaldehyde
Purity 95
Storage temp. Store at -20
Physical Form Liquid-Oil
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2z-3-diethylamino-2-fluoroprop-2-enal

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2z-3-diethylamino-2-fluoroprop-2-enal
pictograms GHS06;GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart