APOH11A9CA71 Apollo Scientific Ltd

2-(Trifluoromethoxy)phenylacetylene

  • MDL number MFCD03839994
  • Cas Number : 886363-40-2
  • Molecular Weight : 186.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethynyl-2-(trifluoromethoxy)benzene
Inchi Code 1S/C9H5F3O/c1-2-7-5-3-4-6-8(7)13-9(10,11)12/h1,3-6H
Inchi Key QWFMOKRCLHCPSJ-UHFFFAOYSA-N

Description

Synonym 1-Ethynyl-2-(trifluoromethoxy)benzene, 2-Ethynylphenyl trifluoromethyl ether
Purity 95%
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethoxyphenylacetylene

Safety Information

signalword Warning
pcodes P210;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethoxyphenylacetylene
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart