APOH11A9CB8C Apollo Scientific Ltd

2,4-Difluorophenylacetone

  • MDL number MFCD02258866
  • Cas Number : 274682-91-6
  • Molecular Weight : 170.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2,4-Difluorophenyl)propan-2-one
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluorophenylacetone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluorophenylacetone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart