APOH11A9CC19 Apollo Scientific Ltd

3-Bromo-2,6-difluorobenzal fluoride

  • Cas Number : 1447671-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-(difluoromethyl)-2,4-difluorobenzene
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=107976

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=107976
Availability details for
Quantity
  • Availability
  • Add to Cart