APOH11A9CC2E Apollo Scientific Ltd

N-Methylperfluorobutanesulfonamide

  • Cas Number : 68298-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Methylnonafluorobutane-1-sulfonamide, 1,1,2,2,3,3,4,4,4-nonafluoro-N-methylbutane-1-sulfonamide
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=109126

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=109126
Availability details for
Quantity
  • Availability
  • Add to Cart