APOH11A9CC34 Apollo Scientific Ltd

3-Fluoro-5-iodo-4-methoxybenzoic acid

  • MDL number MFCD24107165
  • Cas Number : 1542632-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=109320

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=109320
Availability details for
Quantity
  • Availability
  • Add to Cart