APOH11A9CC37 Apollo Scientific Ltd

Potassium 2-(tetrahydro-2h-pyran-2-yloxy)ethyltrifluoroborate

  • MDL number MFCD22684702
  • Cas Number : 1408168-76-2
  • Molecular Weight : 236.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-185
Synonym Potassium trifluoro-[2-(oxan-2-yloxy)ethyl]boranuide
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/potassium-2-tetrahydro-2h-pyran-2-yloxyethyltrifluoroborate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/potassium-2-tetrahydro-2h-pyran-2-yloxyethyltrifluoroborate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart