APOH11A9CCAB Apollo Scientific Ltd

6-Amino-2-fluoro-3-iodobenzonitrile

  • MDL number MFCD09800813
  • Cas Number : 1000577-82-1
  • Molecular Weight : 262.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-2-fluoro-3-iodobenzonitrile
Inchi Code 1S/C7H4FIN2/c8-7-4(3-10)6(11)2-1-5(7)9/h1-2H,11H2
Inchi Key NXYRGTVCZLTGEZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-amino-2-fluoro-3-iodobenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-amino-2-fluoro-3-iodobenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart