APOH11A9CE2A Apollo Scientific Ltd

2-Hydroxychalcone

  • MDL number MFCD00016449
  • Cas Number : 644-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-150
Synonym 3-(2-Hydroxyphenyl)-1-phenylprop-2-en-1-one
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14820

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14820
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart