APOH11A9CEC9 Apollo Scientific Ltd

N,N-Dimethyl-4-[2-(2,4-dinitrophenyl)vinyl]aniline

  • MDL number MFCD00136227
  • Cas Number : 57711-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Dimethylamino)-2',4'-dinitrostilbene, N,N-Dimethyl-4-[2-(2,4-dinitrophenyl)ethenyl]aniline
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16085

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16085
pictograms GHS07
hstatements H302;H315;H317;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart