APOH11A9CF94 Apollo Scientific Ltd

2-Chloro-6-methyl-4-nitrophenol

  • MDL number MFCD00047740
  • Cas Number : 4102-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-2-hydroxy-5-nitrotoluene, 6-Chloro-4-nitro-o-cresol
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=17545

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=17545
Availability details for
Quantity
  • Availability
  • Add to Cart