APOH11A9CFCC Apollo Scientific Ltd

9-Oxodec-2-enoic acid

  • MDL number MFCD00204505
  • Cas Number : 334-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18419

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=18419
Availability details for
Quantity
  • Availability
  • Add to Cart