APOH11A9D074 Apollo Scientific Ltd

Dicyclopropylmethanone

  • MDL number MFCD00001276
  • Cas Number : 1121-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Dicyclopropylketone
Purity 95
Density 0.968
Flash Point 39
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 160-162
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20337

Safety Information

signalword Danger
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20337
pictograms GHS02;GHS05;GHS07
hstatements H226;H302;H315;H318
Availability details for
Quantity
  • Availability
  • Add to Cart