APOH11A9D115 Apollo Scientific Ltd

4-Nitrochalcone

  • MDL number MFCD00007382
  • Cas Number : 1222-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-166
Synonym 3-(4-Nitrophenyl)-1-phenylprop-2-en-1-one, 4-(3-Oxo-3-phenylprop-1-en-1-yl)nitrobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22784

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22784
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart