APOH11A9D143 Apollo Scientific Ltd

4-iodobutyl acetate

  • MDL number MFCD00010664
  • Cas Number : 40596-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23747

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=23747
Availability details for
Quantity
  • Availability
  • Add to Cart