APOH11A9D2BF Apollo Scientific Ltd

4-Hydroxy-3-methoxycinnamic acid

  • MDL number MFCD00004400
  • Cas Number : 1135-24-6
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-173
Synonym 3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, 3-(4-Hydroxy-3-methoxyphenyl)prop-2-enoic acid, Ferulic acid
Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-3-methoxycinnamic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-3-methoxycinnamic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart