APOH11A9D48B Apollo Scientific Ltd

Methyl 4-bromo-3,5-dimethoxybenzoate

  • MDL number MFCD06628784
  • Cas Number : 26050-64-6
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-bromo-3,5-dimethoxybenzoate
Inchi Code 1S/C10H11BrO4/c1-13-7-4-6(10(12)15-3)5-8(14-2)9(7)11/h4-5H,1-3H3
Inchi Key DBPNSECLVZPWET-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-bromo-35-dimethoxybenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-bromo-35-dimethoxybenzoate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart