APOH11A9D595 Apollo Scientific Ltd

5-(Hydroxymethyl)-2-iodophenol

  • MDL number MFCD06202863
  • Cas Number : 773869-57-1
  • Molecular Weight : 250.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 115-118
Iupac Name 5-(hydroxymethyl)-2-iodophenol
Inchi Code 1S/C7H7IO2/c8-6-2-1-5(4-9)3-7(6)10/h1-3,9-10H,4H2
Inchi Key VIWWSAGABDIIFF-UHFFFAOYSA-N

Description

Synonym 3-Hydroxy-4-iodobenzyl alcohol, (3-Hydroxy-4-iodophenyl)methanol
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 115-118
Product Link https://store.apolloscientific.co.uk/product/5-hydroxymethyl-2-iodophenol

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/5-hydroxymethyl-2-iodophenol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart