APOH11A9D5C1 Apollo Scientific Ltd

Iodobenzene dichloride

  • MDL number MFCD00009608
  • Cas Number : 932-72-9
  • Molecular Weight : 274.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 110-112(dec.)
Iupac Name dichloro(phenyl)-lambda~3~-iodane
Inchi Code 1S/C6H5Cl2I/c7-9(8)6-4-2-1-3-5-6/h1-5H
Inchi Key KSRHWBLHVZJTKV-UHFFFAOYSA-N

Description

Purity 70%
Storage temp. Store at -20
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 110-112(dec.)
Product Link https://store.apolloscientific.co.uk/product/iodobenzene-dichloride

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/iodobenzene-dichloride
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart