APOH11A9D714 Apollo Scientific Ltd

4-Phenyl-L-phenylalanine, N-FMOC protected

  • MDL number MFCD00191198
  • Cas Number : 199110-64-0
  • Molecular Weight : 463.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-3-[1,1'-biphenyl]-4-yl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Inchi Code 1S/C30H25NO4/c32-29(33)28(18-20-14-16-22(17-15-20)21-8-2-1-3-9-21)31-30(34)35-19-27-25-12-6-4-10-23(25)24-11-5-7-13-26(24)27/h1-17,27-28H,18-19H2,(H,31,34)(H,32,33)/t28-/m0/s1
Inchi Key VSGACONKQRJFGX-NDEPHWFRSA-N

Description

Synonym (2S)-3-(Biphenyl-4-yl)-2-({[(9H-fluoren-9-y)lmethoxy]carbonyl}amino)propanoic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-phenyl-l-phenylalanine-n-fmoc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-phenyl-l-phenylalanine-n-fmoc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart