APOH11A9D757 Apollo Scientific Ltd

2-(Chloromethyl)-5-methoxy-1,3-benzoxazole

  • MDL number MFCD09836173
  • Cas Number : 63842-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40510

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=40510
Availability details for
Quantity
  • Availability
  • Add to Cart