APOH11A9D758 Apollo Scientific Ltd

Ethyl 4,6-dichloro-5-nitronicotinate

  • MDL number MFCD09842620
  • Cas Number : 154012-15-4
  • Molecular Weight : 265.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-38.5
Synonym Ethyl 4,6-dichloro-5-nitropyridine-3-carboxylate, 2,4-Dichloro-5-(ethoxycarbonyl)-3-nitropyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-46-dichloro-5-nitronicotinate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-46-dichloro-5-nitronicotinate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart