APOH11A9D765 Apollo Scientific Ltd

2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan

  • MDL number MFCD06797643
  • Cas Number : 87776-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(2,3-Dihydrobenzo[b]furan-5-yl)ethan-1-ol
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40494

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=40494
Availability details for
Quantity
  • Availability
  • Add to Cart