APOH11A9D790 Apollo Scientific Ltd

3-Cyano-4-hydroxybenzoic acid

  • MDL number MFCD09998350
  • Cas Number : 70829-28-6
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Carboxy-2-hydroxybenzonitrile, 4-Carboxy-2-cyanophenol
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-cyano-4-hydroxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-cyano-4-hydroxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart