APOH11A9D7D4 Apollo Scientific Ltd

1-Methyl-1H-indole-7-carbonitrile

  • MDL number MFCD09263864
  • Cas Number : 52951-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-Cyano-1-methyl-1H-indole
Purity 97
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42245

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=42245
Availability details for
Quantity
  • Availability
  • Add to Cart