APOH11A9D81C Apollo Scientific Ltd

Methyl 5-bromo-2-hydroxy-3-nitrobenzoate

  • MDL number MFCD00458681
  • Cas Number : 91983-31-2
  • Molecular Weight : 276.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-165
Synonym 5-Bromo-2-hydroxy-3-nitrobenzoic acid methyl ester
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-bromo-2-hydroxy-3-nitrobenzoate

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/methyl-5-bromo-2-hydroxy-3-nitrobenzoate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart