APOH11A9D86D Apollo Scientific Ltd

(S)-1-Methyl-3-(hydroxymethyl)piperidine

  • MDL number MFCD11035736
  • Cas Number : 205194-35-0
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [(3S)-1-methyl-3-piperidinyl]methanol
Inchi Code 1S/C7H15NO/c1-8-4-2-3-7(5-8)6-9/h7,9H,2-6H2,1H3/t7-/m0/s1
Inchi Key UGXQXVDTGJCQHR-ZETCQYMHSA-N

Description

Synonym [(S)-1-Methylpiperidin-3-yl]methanol
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/s-1-methyl-3-hydroxymethylpiperidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/s-1-methyl-3-hydroxymethylpiperidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart