APOH11A9D8CE Apollo Scientific Ltd

2-Iodo-5-nitroanisole

  • MDL number MFCD00024590
  • Cas Number : 5458-84-4
  • Molecular Weight : 279.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-iodo-2-methoxy-4-nitrobenzene
Inchi Code 1S/C7H6INO3/c1-12-7-4-5(9(10)11)2-3-6(7)8/h2-4H,1H3
Inchi Key RBFWMPRFYROKPI-UHFFFAOYSA-N

Description

Synonym 4-Iodo-3-methoxynitrobenzene
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-iodo-5-nitroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-iodo-5-nitroanisole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart