APOH11A9DACF Apollo Scientific Ltd

3-Methyl-1H-indole, N-BOC protected

  • MDL number MFCD13563142
  • Cas Number : 89378-43-8
  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
Iupac Name tert-butyl 3-methyl-1H-indole-1-carboxylate
Inchi Code 1S/C14H17NO2/c1-10-9-15(13(16)17-14(2,3)4)12-8-6-5-7-11(10)12/h5-9H,1-4H3
Inchi Key SMURSSZMVCOKOO-UHFFFAOYSA-N

Description

Melting Point >110
Synonym tert-Butyl 3-methyl-1H-indole-1-carboxylate
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methyl-1h-indole-n-boc-protected

Safety Information

signalword Warning
pcodes P302+P352;P305+P351+P338;P362+P364
MSDS https://store.apolloscientific.co.uk/product/3-methyl-1h-indole-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart