APOH11A9DB65 Apollo Scientific Ltd

2,2'-Bipyridine-5-carboxylic acid

  • MDL number MFCD08234850
  • Cas Number : 1970-80-5
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 196-198
Iupac Name 2,2'-bipyridine-5-carboxylic acid
Inchi Code 1S/C11H8N2O2/c14-11(15)8-4-5-10(13-7-8)9-3-1-2-6-12-9/h1-7H,(H,14,15)
Inchi Key LWMHPOLRJIRLPN-UHFFFAOYSA-N

Description

Synonym 6-(Pyridin-2-yl)nicotinic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 196-198
Product Link https://store.apolloscientific.co.uk/product/22-bipyridine-5-carboxylic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/22-bipyridine-5-carboxylic-acid
pictograms GHS06
hstatements H301;H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart