APOH11A9DBA0 Apollo Scientific Ltd

2-Amino-4-(methoxycarbonyl)benzoic acid

  • MDL number MFCD08690068
  • Cas Number : 85743-02-8
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Carboxy-5-(methoxycarbonyl)aniline
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-methoxycarbonylbenzoic-acid

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-methoxycarbonylbenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart