APOH11A9DBB7 Apollo Scientific Ltd

Ethyl 7-chloroimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD11505035
  • Cas Number : 1204501-38-1
  • Molecular Weight : 224.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-Chloro-2-(ethoxycarbonyl)imidazo[1,2-a]pyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-7-chloroimidazo12-apyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/ethyl-7-chloroimidazo12-apyridine-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart