APOH11A9DBFC Apollo Scientific Ltd

3-Azabicyclo[3.1.0]hexane hydrochloride

  • MDL number MFCD12197361
  • Cas Number : 73799-64-1
  • Molecular Weight : 119.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-162
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-azabicyclo310hexane-hydrochloride

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-azabicyclo310hexane-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart