APOH11A9DC30 Apollo Scientific Ltd

3-Amino-6-bromo-1-methyl-1H-indazole

  • MDL number MFCD11847108
  • Cas Number : 1214899-85-0
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 134-136
Iupac Name 6-bromo-1-methyl-1H-indazol-3-amine
Inchi Code 1S/C8H8BrN3/c1-12-7-4-5(9)2-3-6(7)8(10)11-12/h2-4H,1H3,(H2,10,11)
Inchi Key SJCNMXPRXAGVMK-UHFFFAOYSA-N

Description

Synonym 6-Bromo-1-methyl-1H-indazol-3-amine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 134-136
Product Link https://store.apolloscientific.co.uk/product/3-amino-6-bromo-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-6-bromo-1-methyl-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart