APOH11A9DC5C Apollo Scientific Ltd

2-Methyl-5-nitro-1,3-benzoxazole

  • MDL number MFCD00466748
  • Cas Number : 32046-51-8
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-5-nitro-13-benzoxazole

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-methyl-5-nitro-13-benzoxazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart