APOH11A9DC7B Apollo Scientific Ltd

N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide

  • MDL number MFCD12179438
  • Cas Number : 1157107-85-1
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[(2-Hydroxyethyl)carbamoyl]imidazo[1,2-a]pyridine
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-2-hydroxyethylimidazo12-apyridine-2-carboxamide

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/n-2-hydroxyethylimidazo12-apyridine-2-carboxamide
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart