APOH11A9DEBD Apollo Scientific Ltd

2-(Aminomethyl)pyrimidine hydrochloride

  • MDL number MFCD08062208
  • Cas Number : 372118-67-7
  • Molecular Weight : 145.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-pyrimidinylmethanamine hydrochloride
Inchi Code 1S/C5H7N3.ClH/c6-4-5-7-2-1-3-8-5;/h1-3H,4,6H2;1H
Inchi Key RZNKMZKAXJTLQR-UHFFFAOYSA-N

Description

Synonym (Pyrimidin-2-yl)methylamine hydrochloride, 2-(Aminomethyl)-1,3-diazine hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-aminomethylpyrimidine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminomethylpyrimidine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart