APOH11A9DFCA Apollo Scientific Ltd

5-Chlorothiophene-2-carbonyl chloride

  • MDL number MFCD00043886
  • Cas Number : 42518-98-9
  • Molecular Weight : 181.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 4
Synonym 2-Chloro-5-(chlorocarbonyl)thiophene, 2-Chloro-5-(chloroformyl)thiophene
Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chlorothiophene-2-carbonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-chlorothiophene-2-carbonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart